Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF099067.1 | - | 4 - 5 | 802 | 133 | |
AJ223071.1 | - | 4 - 5 | 892 | 133 | ||
AK078809.1 | - | 4 - 5 | 914 | 133 | ||
AK132803.1 | - | 4 - 5 | 830 | 133 | ||
AK144130.1 | - | 4 - 5 | 776 | 133 | ||
AK147381.1 | - | 4 - 5 | 880 | 133 | ||
AK149584.1 | - | 4 - 5 | 830 | 133 | ||
AK168094.1 | - | 4 - 5 | 813 | 133 | ||
BC057939.1 | - | 4 - 5 | 804 | 5325558 | 133 | |
Y09234.1 | - | 4 - 5 | 892 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |