Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcfl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178254.3 | NP_839985.2 | 1 - 2 | 713 | 141 | |
GenBank mRNA | AK132721.1 | - | 1 - 2 | 370 | 141 | |
AY234363.1 | - | 1 - 2 | 504 | 141 | ||
BC108399.1 | - | 1 - 2 | 713 | 6394526 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |