Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271584.1 | NP_001258513.1 | 1 - 2 | 216 | 124 | |
GenBank mRNA | BC012268.1 | - | 1 - 2 | 3500124 | 126 | |
BY101584.1 | - | 1 - 2 | 151 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |