Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019583.3 | NP_062529.2 | 1 - 2 | 165 | 88 | |
GenBank mRNA | AF208108.1 | - | 1 - 2 | 78 | 88 | |
AF208109.1 | - | 1 - 2 | 78 | 88 | ||
AK045627.1 | - | 1 - 2 | 166 | 88 | ||
AK050139.1 | - | 1 - 2 | 82 | 88 | ||
BC026546.1 | - | 1 - 2 | 65 | 5042466 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |