Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018743.4 | NP_061213.2 | 2 - 3 | 1131 | 88 | |
GenBank mRNA | AF406611.1 | - | 1 - 2 | 642 | 88 | |
AK045235.1 | - | 2 - 3 | 1025 | 88 | ||
AK049675.1 | - | 2 - 3 | 1025 | 88 | ||
AK077561.1 | - | 2 - 3 | 1015 | 88 | ||
AK083589.1 | - | 2 - 3 | 1025 | 88 | ||
AK161270.1 | - | 2 - 3 | 994 | 88 | ||
AK167876.1 | - | 2 - 3 | 1040 | 88 | ||
AK169257.1 | - | 2 - 3 | 1025 | 88 | ||
AK178773.1 | - | 1 - 2 | 120 | 88 | ||
AK188390.1 | - | 1 - 2 | 239 | 88 | ||
AY489184.1 | - | 1 - 2 | 200 | 88 | ||
BC031767.1 | - | 1 - 2 | 485 | 4457846 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |