Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079908.2 | NP_001073377.1 | 12 - 13 | 2414 | 127 | |
NM_001079909.2 | NP_001073378.1 | 11 - 12 | 2153 | 127 | ||
NM_010206.3 | NP_034336.2 | 12 - 13 | 2420 | 127 | ||
GenBank mRNA | AF176552.1 | - | 10 - 11 | 1451 | 127 | |
AK028354.1 | - | 11 - 12 | 1593 | 127 | ||
AK034295.1 | - | 2 - 3 | 240 | 127 | ||
AK049704.1 | - | 12 - 13 | 2380 | 127 | ||
AK167639.1 | - | 3 - 4 | 1717 | 127 | ||
BC010200.1 | - | 12 - 13 | 2313 | 3967874 | 127 | |
BC033447.1 | - | 10 - 11 | 1416 | 4037899 | 127 | |
BC053921.1 | - | 5 - 6 | 1660 | 127 | ||
M28998.1 | - | 11 - 12 | 1718 | 127 | ||
M65053.1 | - | 11 - 12 | 1691 | 127 | ||
U22324.1 | - | 11 - 12 | 1718 | 127 | ||
U23445.1 | - | 10 - 11 | 1451 | 127 | ||
X51893.1 | - | 12 - 13 | 1775 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |