Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145949.2 | NP_666061.3 | 5 - 6 | 535 | 67 | |
GenBank mRNA | AK082385.1 | - | 4 - 5 | 726 | 67 | |
BC026393.1 | - | 5 - 6 | 535 | 4195134 | 67 | |
EF137182.1 | - | 5 - 6 | 535 | 67 | ||
EU440733.1 | - | 4 - 5 | 327 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |