Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025584.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_001025590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_010606.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
NM_010606.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016316.mm.1 |
TC1600002124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600016318.mm.1 |
NM_001025590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016318.mm.1 |
NM_001025590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016318.mm.1 |
TC1600002124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600016314.mm.1 |
NM_001025590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016314.mm.1 |
NM_001025590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016314.mm.1 |
TC1600002124.mm.1 |
MTA TC |