Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133756.4 | NP_598517.1 | 2 - 3 | 213 | 87 | |
GenBank mRNA | AK010393.1 | - | 2 - 3 | 227 | 87 | |
AK077645.1 | - | 2 - 3 | 214 | 87 | ||
AK145957.1 | - | 210 | 87 | |||
AK146686.1 | - | 225 | 87 | |||
BC020174.1 | - | 2 - 3 | 213 | 4505118 | 87 | |
BC096466.1 | - | 2 - 3 | 206 | 3711417 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |