Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 9 - 10 | 1889 | 78 | |
GenBank mRNA | AF093542.1 | - | 10 - 11 | 1123 | 78 | |
AF156934.1 | - | 9 - 10 | 1443 | 78 | ||
AF203445.1 | - | 9 - 10 | 1390 | 78 | ||
AF203446.1 | - | 9 - 10 | 778 | 78 | ||
AF247674.1 | - | 10 - 11 | 1174 | 78 | ||
AK076282.1 | - | 1916 | 78 | |||
AK145556.1 | - | 9 - 10 | 1450 | 78 | ||
AK160291.1 | - | 9 - 10 | 1452 | 78 | ||
AK183147.1 | - | 1 - 2 | 103 | 78 | ||
AK185221.1 | - | 3 - 4 | 215 | 78 | ||
BC003252.1 | - | 9 - 10 | 1446 | 3500413 | 78 | |
BC055071.1 | - | 3865 | 78 | |||
BC066184.1 | - | 9 - 10 | 1425 | 30457053 | 78 | |
BC068314.1 | - | 9 - 10 | 1425 | 30461783 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |