Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159577.1 | NP_001153049.1 | 2 - 3 | 592 | 77 | |
GenBank mRNA | AF034745.1 | - | 2 - 3 | 592 | 77 | |
CA469116.1 | - | 2 - 3 | 629 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |