Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159887.1 | NP_001153359.1 | ||||
NM_001159888.1 | NP_001153360.1 | |||||
NM_001159889.1 | NP_001153361.1 | |||||
NM_023429.4 | NP_075918.1 | |||||
GenBank mRNA | AF295357.1 | - | ||||
AK006013.1 | - | |||||
AK007636.1 | - | |||||
AK043603.1 | - | |||||
AK077904.1 | - | |||||
AK078102.1 | - | |||||
AK078909.1 | - | |||||
AK080795.1 | - | |||||
AK082769.1 | - | |||||
AK082781.1 | - | |||||
AK145907.1 | - | |||||
AK159811.1 | - | |||||
AK167320.1 | - | |||||
AK169715.1 | - | |||||
AY208935.1 | - | |||||
BC006937.1 | - | 2648369 | ||||
CX238085.1 | - | |||||
CX238454.1 | - |
Set Membership: |
Recommended Assay |