Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025834.3 | NP_080110.1 | 1 - 2 | 190 | 82 | |
GenBank mRNA | AK005011.2 | - | 1 - 2 | 134 | 82 | |
AK008819.1 | - | 1 - 2 | 191 | 82 | ||
BC024104.1 | - | 1 - 2 | 165 | 5136392 | 82 | |
BC038045.1 | - | 1 - 2 | 155 | 4196827 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |