Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133962.3 | NP_598723.3 | 12 - 13 | 1768 | 152 | |
GenBank mRNA | AK147923.1 | - | 12 - 13 | 1740 | 152 | |
AK172963.1 | - | 6 - 7 | 871 | 152 | ||
BC060639.1 | - | 12 - 13 | 1740 | 6849875 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |