Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133962.3 | NP_598723.3 | 14 - 15 | 2050 | 57 | |
GenBank mRNA | AK147923.1 | - | 14 - 15 | 2023 | 57 | |
AK172963.1 | - | 8 - 9 | 1153 | 57 | ||
BC034512.1 | - | 3 - 4 | 309 | 57 | ||
BC060639.1 | - | 14 - 15 | 2022 | 6849875 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |