Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019694.1 | NP_062668.1 | 8 - 9 | 1484 | 62 | |
GenBank mRNA | AB041582.1 | - | 8 - 9 | 1497 | 62 | |
AF061026.1 | - | 8 - 9 | 1484 | 62 | ||
AK133849.1 | - | 8 - 9 | 1504 | 62 | ||
AK161638.1 | - | 2 - 3 | 1206 | 62 | ||
AK218029.1 | - | 3 - 4 | 326 | 62 | ||
BC023862.1 | - | 8 - 9 | 1481 | 62 | ||
BC046326.1 | - | 8 - 9 | 1491 | 62 | ||
BC055865.1 | - | 8 - 9 | 1502 | 62 | ||
BC061115.1 | - | 8 - 9 | 1497 | 6533034 | 62 | |
BC061690.1 | - | 8 - 9 | 1484 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |