Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145858.1 | NP_001139330.1 | 11 - 12 | 1707 | 126 | |
NM_001145859.1 | NP_001139331.1 | 11 - 12 | 1710 | 126 | ||
NM_011893.3 | NP_036023.3 | 11 - 12 | 1607 | 126 | ||
GenBank mRNA | AK089455.1 | - | 11 - 12 | 1538 | 126 | |
AK143548.1 | - | 11 - 12 | 1712 | 126 | ||
AK150265.1 | - | 11 - 12 | 1709 | 126 | ||
AK155103.1 | - | 11 - 12 | 1564 | 126 | ||
AK155898.1 | - | 11 - 12 | 1607 | 126 | ||
AK155970.1 | - | 11 - 12 | 1655 | 126 | ||
BC010198.1 | - | 11 - 12 | 1545 | 3967543 | 126 | |
BC085497.1 | - | 11 - 12 | 1598 | 5696296 | 126 | |
L14543.1 | - | 11 - 12 | 1541 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |