Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009788.4 | NP_033918.1 | 9 - 10 | 739 | 139 | |
GenBank mRNA | AK002635.1 | - | 9 - 10 | 741 | 139 | |
AK005081.1 | - | 9 - 10 | 738 | 139 | ||
AK005243.1 | - | 9 - 10 | 739 | 139 | ||
AK028050.1 | - | 9 - 10 | 740 | 139 | ||
AK028327.1 | - | 9 - 10 | 738 | 139 | ||
AK048517.1 | - | 9 - 10 | 740 | 139 | ||
AK077499.1 | - | 9 - 10 | 738 | 139 | ||
AK078358.1 | - | 9 - 10 | 737 | 139 | ||
BC016421.1 | - | 9 - 10 | 670 | 4527000 | 139 | |
M21531.1 | - | 9 - 10 | 719 | 139 | ||
M23663.1 | - | 6 - 7 | 301 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |