Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111096.1 | NP_001104566.1 | 11 - 12 | 1456 | 98 | |
NM_010747.2 | NP_034877.2 | 11 - 12 | 1393 | 98 | ||
GenBank mRNA | AK028112.1 | - | 11 - 12 | 1389 | 98 | |
AK151914.1 | - | 11 - 12 | 1452 | 98 | ||
AK152199.1 | - | 11 - 12 | 1454 | 98 | ||
AK153038.1 | - | 11 - 12 | 1390 | 98 | ||
AK170561.1 | - | 11 - 12 | 1454 | 98 | ||
BC003253.1 | - | 11 - 12 | 1333 | 3500419 | 98 | |
BC031547.1 | - | 11 - 12 | 1426 | 3589207 | 98 | |
BC157998.1 | - | 11 - 12 | 1225 | 9007404 | 98 | |
M64608.1 | - | 11 - 12 | 1442 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |