Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081205.1 | NP_001074674.1 | 2 - 3 | 409 | 63 | |
GenBank mRNA | AK014427.1 | - | 2 - 3 | 362 | 63 | |
AK075654.1 | - | 2 - 3 | 410 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |