Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB047647.1 | - | 5 - 6 | 572 | 102 | |
AB052779.1 | - | 7 - 8 | 829 | 102 | ||
AB182651.1 | - | 7 - 8 | 947 | 102 | ||
AB182652.1 | - | 5 - 6 | 572 | 102 | ||
AF287260.1 | - | 5 - 6 | 710 | 102 | ||
AK005918.1 | - | 5 - 6 | 729 | 102 | ||
AK019928.1 | - | 7 - 8 | 947 | 102 | ||
AK088919.1 | - | 7 - 8 | 846 | 102 | ||
AY049978.1 | - | 7 - 8 | 868 | 102 | ||
BC022605.1 | - | 7 - 8 | 803 | 4164994 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |