Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205253.1 | NP_001192182.1 | 2 - 3 | 1085 | 144 | |
NM_019576.2 | NP_062522.1 | 3 - 4 | 1228 | 144 | ||
GenBank mRNA | AB039946.1 | - | 2 - 3 | 1056 | 144 | |
AK049636.1 | - | 3 - 4 | 1191 | 144 | ||
AK052555.1 | - | 2 - 3 | 1026 | 144 | ||
BC119603.2 | - | 2 - 3 | 1056 | 40060210 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |