Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172526.3 | NP_766114.1 | 8 - 9 | 1463 | 66 | |
GenBank mRNA | AK039884.1 | - | 8 - 9 | 1464 | 66 | |
AK076743.1 | - | 3 - 4 | 279 | 66 | ||
BC055114.1 | - | 3 - 4 | 278 | 66 | ||
BC147554.1 | - | 7 - 8 | 1167 | 9056545 | 66 | |
BC147555.1 | - | 7 - 8 | 1167 | 9056546 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |