Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnisr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025669.1 | NP_079945.1 | 5 - 6 | 1243 | 99 | |
GenBank mRNA | AK012036.1 | - | 5 - 6 | 785 | 99 | |
AK017507.1 | - | 5 - 6 | 770 | 99 | ||
AK031964.1 | - | 5 - 6 | 779 | 99 | ||
AK077040.1 | - | 3 - 4 | 2479 | 99 | ||
AK083021.1 | - | 5 - 6 | 769 | 99 | ||
AK160288.1 | - | 5 - 6 | 769 | 99 | ||
AK162695.1 | - | 5 - 6 | 738 | 99 | ||
BC060111.1 | - | 5 | 695 | 160 | ||
BC141031.1 | - | 5 - 6 | 1107 | 9055643 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |