Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024434.6 | NP_077754.3 | 2 - 3 | 298 | 99 | |
GenBank mRNA | AF334160.1 | - | 2 - 3 | 259 | 99 | |
AK002819.1 | - | 2 - 3 | 234 | 99 | ||
AK005334.1 | - | 2 - 3 | 211 | 99 | ||
AK008600.1 | - | 2 - 3 | 185 | 99 | ||
AK010384.1 | - | 2 - 3 | 184 | 99 | ||
AK010502.1 | - | 2 - 3 | 251 | 99 | ||
AK011604.1 | - | 2 - 3 | 184 | 99 | ||
AK089023.1 | - | 2 - 3 | 265 | 99 | ||
AK166958.1 | - | 2 - 3 | 182 | 99 | ||
AK169032.1 | - | 2 - 3 | 184 | 99 | ||
BC016536.1 | - | 2 - 3 | 298 | 3488469 | 99 | |
CT010237.1 | - | 2 - 3 | 212 | 99 | ||
CT010301.1 | - | 2 - 3 | 212 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |