Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021896.5 | NP_068696.2 | 3 - 4 | 627 | 94 | |
GenBank mRNA | AF297082.1 | - | 3 - 4 | 602 | 94 | |
AK004815.1 | - | 3 - 4 | 561 | 94 | ||
AK028558.1 | - | 3 - 4 | 550 | 94 | ||
AK031305.1 | - | 3 - 4 | 540 | 94 | ||
BC094310.1 | - | 3 - 4 | 683 | 6838836 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |