Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026460.3 | NP_080736.2 | 3 - 4 | 525 | 85 | |
GenBank mRNA | AF251276.1 | - | 3 - 4 | 510 | 85 | |
AK007347.1 | - | 3 - 4 | 526 | 85 | ||
AK007510.1 | - | 3 - 4 | 500 | 85 | ||
AK008931.1 | - | 3 - 4 | 527 | 85 | ||
BC098466.1 | - | 3 - 4 | 502 | 6433250 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |