Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153135.3 | NP_694775.1 | 13 - 14 | 2239 | 74 | |
GenBank mRNA | AJ487854.1 | - | 13 - 14 | 2240 | 74 | |
AK220417.1 | - | 3 - 4 | 580 | 74 | ||
BC137615.1 | - | 13 - 14 | 2030 | 8860634 | 74 | |
BC144783.1 | - | 11 - 12 | 1847 | 9053331 | 74 | |
BC144784.1 | - | 11 - 12 | 1808 | 9053332 | 74 | |
BC145695.1 | - | 10 - 11 | 1814 | 40130998 | 74 | |
FJ824810.1 | - | 12 - 13 | 2118 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |