Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172808.2 | NP_766396.2 | 9 - 10 | 988 | 145 | |
GenBank mRNA | AK077206.1 | - | 9 - 10 | 989 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |