Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194055.3 | NP_918944.2 | 6 - 7 | 988 | 101 | |
GenBank mRNA | AK042164.1 | - | 6 - 7 | 600 | 101 | |
BC031468.1 | - | 4 - 5 | 291 | 4037840 | 101 | |
BC059280.1 | - | 4 - 5 | 289 | 101 | ||
DV659240.1 | - | 4 - 5 | 344 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |