Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400002069.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002069.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002069.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002071.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002071.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002071.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002072.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002072.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002072.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002075.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002075.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002075.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002070.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002070.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002070.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002073.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002073.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002073.mm.1 |
TC1400000263.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002068.mm.1 |
NM_007479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002068.mm.1 |
NM_007479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002068.mm.1 |
TC1400000263.mm.1 |
MTA TC |