Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181584.4 | NP_853615.1 | 1 - 2 | 241 | 80 | |
GenBank mRNA | AK031227.1 | - | 1 - 2 | 239 | 80 | |
AY057988.1 | - | 1 - 2 | 127 | 80 | ||
BC141240.1 | - | 1 - 2 | 121 | 9055852 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |