Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177680.3 | NP_808348.2 | 2 - 3 | 206 | 113 | |
GenBank mRNA | AK135309.1 | - | 2 - 3 | 463 | 113 | |
BC044650.1 | - | 1 - 2 | 78 | 113 | ||
CN528111.1 | - | 2 - 3 | 128 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |