Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007426.4 | NP_031452.2 | 3 - 4 | 849 | 85 | |
GenBank mRNA | AF004326.1 | - | 3 - 4 | 775 | 85 | |
AK019860.1 | - | 3 - 4 | 838 | 85 | ||
AK048622.1 | - | 3 - 4 | 806 | 85 | ||
AK143974.1 | - | 3 - 4 | 837 | 85 | ||
AK156132.1 | - | 3 - 4 | 820 | 85 | ||
BC027216.1 | - | 3 - 4 | 802 | 3494566 | 85 | |
BC138309.1 | - | 3 - 4 | 743 | 8861329 | 85 | |
BC138313.1 | - | 3 - 4 | 743 | 8861333 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |