Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011369.2 | NP_035499.1 | 6 - 7 | 923 | 154 | |
GenBank mRNA | AF017152.1 | - | 6 - 7 | 923 | 154 | |
AK137496.1 | - | 6 - 7 | 939 | 154 | ||
AK144966.1 | - | 6 - 7 | 946 | 154 | ||
AK149580.1 | - | 6 - 7 | 940 | 154 | ||
AK162943.1 | - | 6 - 7 | 937 | 154 | ||
BC070455.1 | - | 6 - 7 | 943 | 30606785 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |