Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110809.2 | NP_001104279.1 | 4 - 5 | 374 | 71 | |
NM_001286857.1 | NP_001273786.1 | 4 - 5 | 988 | 71 | ||
NM_025663.4 | NP_079939.3 | 5 - 6 | 509 | 71 | ||
GenBank mRNA | AK011617.1 | - | 4 - 5 | 373 | 71 | |
AK017310.1 | - | 4 - 5 | 324 | 71 | ||
AK018306.1 | - | 4 - 5 | 348 | 71 | ||
AK078828.1 | - | 4 - 5 | 372 | 71 | ||
AK169111.1 | - | 4 - 5 | 363 | 71 | ||
BC002265.1 | - | 4 - 5 | 345 | 71 | ||
BC019490.1 | - | 5 - 6 | 491 | 4039649 | 71 | |
BC080676.1 | - | 4 - 5 | 966 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |