Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286538.1 | NP_001273467.1 | 2 - 3 | 457 | 80 | |
NM_001286540.1 | NP_001273469.1 | 2 - 3 | 457 | 80 | ||
NM_011014.3 | NP_035144.1 | 2 - 3 | 457 | 80 | ||
GenBank mRNA | AF004927.1 | - | 2 - 3 | 411 | 80 | |
AF030198.1 | - | 2 - 3 | 371 | 80 | ||
AK002718.1 | - | 2 - 3 | 390 | 80 | ||
AK154300.1 | - | 2 - 3 | 429 | 80 | ||
AK159886.1 | - | 2 - 3 | 391 | 80 | ||
AY390765.1 | - | 2 - 3 | 350 | 80 | ||
AY390766.1 | - | 2 - 3 | 350 | 80 | ||
BC002000.1 | - | 2 - 3 | 406 | 3484605 | 80 | |
BC019930.1 | - | 2 - 3 | 385 | 5123891 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |