Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166493.1 | NP_001159965.1 | 9 - 10 | 1485 | 99 | |
NM_207246.4 | NP_997129.1 | 10 - 11 | 1477 | 99 | ||
GenBank mRNA | AB154824.1 | - | 8 - 9 | 1172 | 99 | |
AK173041.1 | - | 9 - 10 | 1370 | 99 | ||
BC066069.1 | - | 8 - 9 | 1115 | 6851971 | 99 | |
BC066127.1 | - | 10 - 11 | 1402 | 30280332 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |