Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018861.3 | NP_061349.3 | 1 - 2 | 769 | 81 | |
GenBank mRNA | AF246129.1 | - | 1 - 2 | 528 | 81 | |
AK044280.1 | - | 1 - 2 | 740 | 81 | ||
AK136900.1 | - | 1 - 2 | 769 | 81 | ||
AK139248.1 | - | 1 - 2 | 768 | 81 | ||
AK140866.1 | - | 1 - 2 | 770 | 81 | ||
AK148075.1 | - | 1 - 2 | 770 | 81 | ||
BC043483.1 | - | 1 - 2 | 727 | 5003744 | 81 | |
BC052733.1 | - | 1 - 2 | 643 | 6816689 | 81 | |
U75215.1 | - | 1 - 2 | 639 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |