Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031848.1 | - | 6 - 7 | 949 | 84 | |
AK036017.1 | - | 6 - 7 | 886 | 84 | ||
AK041354.1 | - | 7 - 8 | 1052 | 84 | ||
AK042540.1 | - | 6 - 7 | 880 | 84 | ||
AY753561.1 | - | 6 - 7 | 842 | 84 | ||
AY753562.1 | - | 6 - 7 | 842 | 84 | ||
BC056428.1 | - | 6 - 7 | 873 | 5716245 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |