Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175265.4 | NP_780474.1 | 8 - 9 | 996 | 145 | |
GenBank mRNA | AK034930.1 | - | 8 - 9 | 997 | 145 | |
AK047285.1 | - | 2 - 3 | 198 | 145 | ||
AK141389.1 | - | 8 - 9 | 947 | 145 | ||
AK167430.1 | - | 8 - 9 | 928 | 145 | ||
BC052183.1 | - | 8 - 9 | 897 | 6336038 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |