Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134426.1 | NP_001127898.1 | 2 - 3 | 803 | 97 | |
NM_001134427.1 | NP_001127899.1 | 2 - 3 | 593 | 97 | ||
NM_175565.3 | NP_780774.1 | 2 - 3 | 803 | 97 | ||
NM_175833.2 | NP_787027.1 | 2 - 3 | 803 | 97 | ||
GenBank mRNA | AB046372.1 | - | 2 - 3 | 391 | 97 | |
AF215660.1 | - | 2 - 3 | 561 | 97 | ||
AF320340.1 | - | 2 - 3 | 561 | 97 | ||
AK172053.1 | - | 2 - 3 | 512 | 97 | ||
BC031362.1 | - | 2 - 3 | 593 | 97 | ||
BC052770.1 | - | 2 - 3 | 697 | 6306557 | 97 | |
BC062156.1 | - | 3 - 4 | 649 | 6507616 | 97 | |
BC096479.1 | - | 2 - 3 | 654 | 4979176 | 97 | |
L22067.1 | - | 2 - 3 | 644 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |