Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028138.1 | NP_082414.1 | 7 - 8 | 983 | 72 | |
GenBank mRNA | AK012806.1 | - | 7 - 8 | 966 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |