Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177290.3 | NP_796264.2 | 4 - 5 | 1115 | 84 | |
GenBank mRNA | AK029281.1 | - | 4 - 5 | 1521 | 84 | |
AK053056.1 | - | 1640 | 84 | |||
AK082444.1 | - | 1650 | 84 | |||
BC120691.1 | - | 4 - 5 | 1115 | 40129614 | 84 | |
BC125343.1 | - | 4 - 5 | 1115 | 40129858 | 84 | |
BC144785.1 | - | 4 - 5 | 1120 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |