Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010250.5 | NP_034380.1 | 3 - 4 | 962 | 73 | |
GenBank mRNA | AK034095.1 | - | 3 - 4 | 963 | 73 | |
AK039496.1 | - | 3 - 4 | 246 | 73 | ||
AK141596.1 | - | 3 - 4 | 822 | 73 | ||
BC132331.1 | - | 3 - 4 | 638 | 40130608 | 73 | |
BC137733.1 | - | 3 - 4 | 641 | 8860752 | 73 | |
M63436.1 | - | 3 - 4 | 635 | 73 | ||
M86566.1 | - | 3 - 4 | 330 | 73 | ||
X61430.1 | - | 2 - 3 | 218 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |