Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081136.1 | NP_001074605.1 | 3 - 4 | 343 | 82 | |
GenBank mRNA | AK137763.1 | - | 3 - 4 | 340 | 82 | |
BC147680.1 | - | 3 - 4 | 314 | 9056653 | 82 | |
BC147683.1 | - | 3 - 4 | 314 | 9056656 | 82 | |
BC147829.1 | - | 3 - 4 | 272 | 9087546 | 82 | |
BC147832.1 | - | 3 - 4 | 272 | 9087549 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |