Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700011508.mm.1 |
NM_001114361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011508.mm.1 |
NM_001114361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011508.mm.1 |
TC1700001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_001114361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_001114361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_001114362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_001114362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_001286567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_199466.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
NM_199466.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011505.mm.1 |
TC1700001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_001114361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_001114361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_001114362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_001114362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_001286567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_199466.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
NM_199466.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011506.mm.1 |
TC1700001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011504.mm.1 |
NM_001286567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011504.mm.1 |
TC1700001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_001114361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_001114361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_001114362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_001114362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_001286567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_199466.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
NM_199466.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011503.mm.1 |
TC1700001241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_001114361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_001114361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_001114362.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_001114362.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_001286567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_199466.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
NM_199466.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011499.mm.1 |
TC1700001241.mm.1 |
MTA TC |