Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003919.1 | NP_001003919.1 | 2 - 3 | 322 | 70 | |
GenBank mRNA | AK049419.1 | - | 2 - 3 | 254 | 70 | |
AK076357.1 | - | 2 - 3 | 273 | 70 | ||
AK152868.1 | - | 2 - 3 | 231 | 70 | ||
BC079656.1 | - | 2 - 3 | 322 | 6854039 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |