Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039551.1 | NP_001034640.1 | 5 - 6 | 1837 | 78 | |
NM_053186.2 | NP_444416.2 | 5 - 6 | 1837 | 78 | ||
GenBank mRNA | AF216964.1 | - | 5 - 6 | 906 | 78 | |
AK143035.1 | - | 3 - 4 | 369 | 78 | ||
BC052714.1 | - | 5 - 6 | 904 | 78 | ||
BC080275.1 | - | 5 - 6 | 1264 | 78 | ||
BC108417.1 | - | 5 - 6 | 1091 | 78 | ||
BC132285.1 | - | 5 - 6 | 889 | 78 | ||
BC144720.1 | - | 5 - 6 | 888 | 78 | ||
GQ225746.2 | - | 5 - 6 | 823 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |