Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010830.2 | NP_034960.1 | 6 - 7 | 3651 | 80 | |
GenBank mRNA | AK010643.1 | - | 3 - 4 | 812 | 80 | |
AK088259.1 | - | 6 - 7 | 3640 | 80 | ||
BC035274.1 | - | 4 - 5 | 3049 | 80 | ||
BC051160.1 | - | 6 - 7 | 3596 | 4953238 | 80 | |
BC051634.1 | - | 6 - 7 | 3606 | 6518577 | 80 | |
U42190.1 | - | 6 - 7 | 3627 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |